Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITFG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001305002.1 | NP_001291931.1 | 6 | 541 | ||
NM_030790.4 | NP_110417.2 | 6 | 818 | |||
GenBank mRNA | AF212247.1 | - | 6 | 726 | ||
AF503339.1 | - | 6 | 608 | |||
AK027596.1 | - | 6 | 609 | |||
AK301908.1 | - | 6 | 493 | |||
BC006321.2 | - | 6 | 591 | 4137934 | ||
BC024162.1 | - | 6 | 652 | 4046297 |