Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030891.4 | NP_112153.1 | 2 | 1605 | ||
GenBank mRNA | AK127604.1 | - | 3 | 1222 | ||
AY358120.1 | - | 2 | 1567 | |||
BC064321.1 | - | 2 | 1605 |