Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM117A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030802.3 | NP_110429.1 | 8 | 1183 | ||
XM_017025182.1 | XP_016880671.1 | 8 | 1941 | |||
GenBank mRNA | AF123073.1 | - | 8 | 1144 | ||
AK302376.1 | - | 8 | 997 | |||
BC037572.1 | - | 8 | 1195 | 5526752 | ||
BC065199.1 | - | 8 | 1148 | 5214423 |