Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMAN2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142292.1 | NP_001135764.1 | 4 | 508 | ||
NM_001322347.1 | NP_001309276.1 | 3 | 405 | |||
NM_001322350.1 | NP_001309279.1 | 3 | 405 | |||
NM_001322352.1 | NP_001309281.1 | 5 | 605 | |||
NM_030805.3 | NP_110432.1 | 4 | 508 | |||
GenBank mRNA | AJ549957.1 | - | 4 | 472 | ||
AK075347.1 | - | 4 | 496 | |||
AK222828.1 | - | 4 | 486 | |||
AK295463.1 | - | 3 | 380 | |||
AK304515.1 | - | 3 | 376 | |||
AK308085.1 | - | 4 | 495 | |||
AK310100.1 | - | 4 | 525 | |||
AK312249.1 | - | 4 | 501 | |||
AL136617.1 | - | 4 | 500 | |||
AY358929.1 | - | 4 | 479 | |||
BC000347.2 | - | 4 | 471 | |||
BC005822.2 | - | 4 | 471 | |||
BC005862.2 | - | 4 | 471 | |||
BC067265.1 | - | 4 | 478 | 6502212 | ||
BX648002.1 | - | 4 | 486 | |||
DC354352.1 | - | 4 | 510 |