Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXNDC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145549.2 | NP_001139021.1 | 9 | 1093 | ||
NM_030810.3 | NP_110437.2 | 9 | 1129 | |||
GenBank mRNA | AJ440721.1 | - | 9 | 884 | ||
AK025006.1 | - | 3 | 869 | |||
AK075291.1 | - | 9 | 1103 | |||
AK098089.1 | - | 12 | 1105 | |||
AK315598.1 | - | 9 | 1094 | |||
AL833008.1 | - | 9 | 973 | |||
AL834423.1 | - | 6 | 527 | |||
AY326464.1 | - | 9 | 1117 | |||
AY358646.1 | - | 9 | 1123 | |||
BC001199.1 | - | 9 | 848 | 3356029 |
Gene Symbol: BLOC1S5-TXNDC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037616.1 | 12 | 1250 | |||
GenBank mRNA | BC052310.1 | - | 12 | 1200 | 6671945 |