Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD13C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030816.4 | NP_110443.3 | 12 | 1765 | ||
XM_006710929.3 | XP_006710992.1 | 12 | 1778 | |||
XM_017002413.1 | XP_016857902.1 | 10 | 988 | |||
XM_017002414.1 | XP_016857903.1 | 11 | 1070 | |||
XM_017002415.1 | XP_016857904.1 | 11 | 1049 | |||
XM_017002416.1 | XP_016857905.1 | 7 | 734 | |||
GenBank mRNA | AK074187.1 | - | 4 | 389 | ||
AL136717.1 | - | 11 | 1661 | |||
BC028840.1 | - | 12 | 1754 | 4558748 |