Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GFOD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030819.3 | NP_110446.3 | 3 | 569 | ||
NR_027398.1 | 2 | 223 | ||||
XM_006721288.3 | XP_006721351.1 | 3 | 312 | |||
XM_017023738.1 | XP_016879227.1 | 3 | 519 | |||
GenBank mRNA | AK074382.1 | - | 4 | 522 | ||
AK091606.1 | - | 1 | 1446 | |||
AK225997.1 | - | 3 | 539 | |||
AY358169.1 | - | 2 | 279 | |||
BC000757.2 | - | 2 | 176 | 2820364 | ||
BC002943.1 | - | 2 | 196 | |||
BC107585.1 | - | 3 | 519 | 6463662 | ||
BI826603.1 | - | 3 | 622 |