Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDADC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193478.1 | NP_001180407.1 | 3 | 363 | ||
NM_030911.3 | NP_112173.1 | 3 | 363 | |||
NR_036439.1 | 3 | 363 | ||||
XM_011535250.2 | XP_011533552.1 | 3 | 327 | |||
XM_011535251.1 | XP_011533553.1 | 3 | 307 | |||
XM_011535253.1 | XP_011533555.1 | 3 | 399 | |||
XM_017020778.1 | XP_016876267.1 | 1 | 46 | |||
XM_017020779.1 | XP_016876268.1 | 3 | 355 | |||
XM_017020780.1 | XP_016876269.1 | 3 | 705 | |||
XM_017020781.1 | XP_016876270.1 | 3 | 355 | |||
XM_017020782.1 | XP_016876271.1 | 2 | 153 | |||
XM_017020783.1 | XP_016876272.1 | 3 | 705 | |||
GenBank mRNA | AK297400.1 | - | 3 | 327 | ||
AK312835.1 | - | 3 | 333 | |||
AY027525.1 | - | 3 | 312 | |||
BC009562.1 | - | 3 | 327 | |||
BC032889.1 | - | 3 | 338 | |||
BC048092.1 | - | 3 | 299 | |||
BC070238.1 | - | 3 | 331 | |||
BC092434.1 | - | 3 | 341 | |||
BC125202.1 | - | 3 | 277 | 40123430 | ||
DA400049.1 | - | 3 | 363 | |||
DA560379.1 | - | 3 | 363 |