Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDADC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193478.1 | NP_001180407.1 | 1, 2 | 207 | ||
NM_030911.3 | NP_112173.1 | 1, 2 | 207 | |||
NR_036439.1 | 1, 2 | 207 | ||||
XM_011535253.1 | XP_011533555.1 | 1, 2 | 243 | |||
XM_017020779.1 | XP_016876268.1 | 1, 2 | 199 | |||
XM_017020781.1 | XP_016876270.1 | 1, 2 | 199 | |||
GenBank mRNA | AK297400.1 | - | 1, 2 | 171 | ||
AK312835.1 | - | 1, 2 | 177 | |||
AY027525.1 | - | 1, 2 | 156 | |||
BC009562.1 | - | 1, 2 | 171 | |||
BC032889.1 | - | 1, 2 | 182 | |||
BC048092.1 | - | 1, 2 | 143 | |||
BC070238.1 | - | 1, 2 | 175 | |||
BC092434.1 | - | 1, 2 | 185 | |||
BC125202.1 | - | 1, 2 | 121 | 40123430 | ||
DA400049.1 | - | 1, 2 | 207 | |||
DA560379.1 | - | 1, 2 | 207 |