Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030912.2 | NP_112174.2 | 1 | 346 | ||
GenBank mRNA | AF220034.1 | - | 1 | 223 | ||
AF281046.1 | - | 1 | 232 | |||
AL360160.1 | - | 1 | 232 | |||
BC021925.1 | - | 1 | 345 | 4857393 | ||
BG746631.1 | - | 1 | 346 |