Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CX3CR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171171.1 | NP_001164642.1 | 2 | 870 | ||
NM_001171172.1 | NP_001164643.1 | 2 | 830 | |||
NM_001171174.1 | NP_001164645.1 | 2 | 766 | |||
NM_001337.3 | NP_001328.1 | 2 | 722 | |||
GenBank mRNA | AK225764.1 | - | 2 | 723 | ||
AK307911.1 | - | 3 | 522 | |||
AK312373.1 | - | 2 | 721 | |||
BC028078.1 | - | 2 | 706 | 5216452 | ||
U20350.1 | - | 2 | 717 | |||
U28934.1 | - | 2 | 675 |