Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KAZALD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030929.4 | NP_112191.2 | 5 | 1998 | ||
GenBank mRNA | AK172862.1 | - | 5 | 1249 | ||
AK225949.1 | - | 5 | 1250 | |||
AY359087.1 | - | 5 | 1254 | |||
BC007758.2 | - | 5 | 1399 | 4122366 | ||
BI766345.1 | - | 4 | 710 |