Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEFB126 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030931.3 | NP_112193.1 | 1, 2 | 142 | ||
GenBank mRNA | AF525928.1 | - | 1, 2 | 66 | ||
AJ236910.1 | - | 1, 2 | 84 | |||
AK225987.1 | - | 1, 2 | 105 | |||
AY122478.1 | - | 1, 2 | 60 | |||
BX108307.1 | - | 1, 2 | 325 |