Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIAPH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042517.1 | NP_001035982.1 | 12 | 1476 | ||
NM_001258366.1 | NP_001245295.1 | 11 | 1443 | |||
NM_001258367.1 | NP_001245296.1 | 10 | 1338 | |||
NM_001258368.1 | NP_001245297.1 | 10 | 1266 | |||
NM_001258369.1 | NP_001245298.1 | 12 | 1476 | |||
NM_001258370.1 | NP_001245299.1 | 6 | 647 | |||
NM_030932.3 | NP_112194.2 | 6 | 647 | |||
XM_006719876.1 | XP_006719939.1 | 6 | 627 | |||
XM_011535258.2 | XP_011533560.1 | 12 | 1419 | |||
XM_011535263.2 | XP_011533565.1 | 12 | 1419 | |||
XM_011535265.2 | XP_011533567.1 | 12 | 1419 | |||
XM_017020789.1 | XP_016876278.1 | 12 | 1419 | |||
GenBank mRNA | AB244756.1 | - | 11 | 1224 | ||
AB244757.1 | - | 10 | 1119 | |||
AB244758.1 | - | 10 | 1047 | |||
AK092024.1 | - | 13 | 1398 | |||
AK298509.1 | - | 12 | 1262 | |||
AK307652.1 | - | 6 | 627 | |||
AK316474.1 | - | 12 | 1389 | |||
AL137718.1 | - | 6 | 619 | |||
AY750055.1 | - | 12 | 1349 | |||
AY818645.1 | - | 12 | 1257 | |||
BC034952.1 | - | 6 | 651 | 4830888 | ||
BC048963.1 | - | 6 | 603 | 5265490 | ||
BC068504.1 | - | 6 | 603 | 4838507 | ||
BX649186.1 | - | 12 | 1418 |