Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHCBP1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030933.2 | NP_112195.2 | 5 | 1234 | ||
GenBank mRNA | AF288397.1 | - | 6 | 892 | ||
AF288398.1 | - | 5 | 1233 | |||
BC026084.1 | - | 4 | 1414 | |||
BC050305.1 | - | 5 | 1186 | 4837018 | ||
BC132764.1 | - | 5 | 1210 | 40146786 | ||
BC144326.1 | - | 5 | 1206 |