Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSC22D4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303043.1 | NP_001289972.1 | 5 | 2212 | ||
NM_030935.4 | NP_112197.1 | 5 | 2315 | |||
NR_130118.1 | 5 | 1470 | ||||
NR_130119.1 | 5 | 1467 | ||||
GenBank mRNA | AJ133115.1 | - | 5 | 1900 | ||
BC001486.2 | - | 5 | 2315 | 2966929 | ||
BC001966.1 | - | 5 | 2168 | 3457297 | ||
BC002972.1 | - | 5 | 2222 | 3538175 | ||
BC010406.2 | - | 5 | 2229 | 3832780 |