Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039577.3 | NP_001034666.1 | 4 | 535 | ||
NM_001320153.1 | NP_001307082.1 | 4 | 535 | |||
NM_001320155.1 | NP_001307084.1 | 4 | 535 | |||
NM_030937.4 | NP_112199.2 | 4 | 535 | |||
NR_135154.1 | 4 | 535 | ||||
XM_011542216.2 | XP_011540518.1 | 4 | 610 | |||
XM_011542219.2 | XP_011540521.1 | 2 | 126 | |||
XM_011542221.2 | XP_011540523.1 | 2 | 125 | |||
XM_017002420.1 | XP_016857909.1 | 4 | 533 | |||
XM_017002421.1 | XP_016857910.1 | 4 | 519 | |||
XM_017002422.1 | XP_016857911.1 | 4 | 532 | |||
XM_017002423.1 | XP_016857912.1 | 4 | 610 | |||
GenBank mRNA | AF251294.1 | - | 2 | 171 | ||
AK057133.1 | - | 2 | 125 | |||
AK074112.1 | - | 4 | 506 | |||
AK225758.1 | - | 4 | 530 | |||
AK293451.1 | - | 1 | 13 | |||
AL834441.1 | - | 4 | 466 | |||
AY037150.1 | - | 4 | 509 | |||
AY116620.1 | - | 4 | 524 | |||
BC016333.1 | - | 4 | 515 | |||
BM153748.1 | - | 4 | 535 | |||
DA360436.1 | - | 4 | 534 |