Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ISCA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030940.3 | NP_112202.2 | 4 | 775 | ||
GenBank mRNA | AF038186.1 | - | 1 | 190 | ||
AF284752.1 | - | 4 | 730 | |||
AK055643.1 | - | 4 | 738 | |||
AK225937.1 | - | 4 | 738 | |||
AK296093.1 | - | 4 | 961 | |||
AL834356.1 | - | 4 | 720 | |||
BC002675.2 | - | 4 | 729 | 3608301 | ||
BC053900.1 | - | 4 | 696 | |||
BC071621.1 | - | 4 | 709 | 5107151 |