Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOCK8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190458.1 | NP_001177387.1 | 3 | 291 | ||
NM_001193536.1 | NP_001180465.1 | 3 | 291 | |||
NM_203447.3 | NP_982272.2 | 4 | 506 | |||
XM_011518045.2 | XP_011516347.1 | 4 | 361 | |||
XM_011518046.2 | XP_011516348.1 | 4 | 404 | |||
XM_011518047.2 | XP_011516349.1 | 4 | 587 | |||
XM_011518048.2 | XP_011516350.1 | 3 | 326 | |||
XM_017015173.1 | XP_016870662.1 | 4 | 361 | |||
XM_017015174.1 | XP_016870663.1 | 4 | 441 | |||
GenBank mRNA | AB191037.1 | - | 4 | 506 | ||
AK093572.1 | - | 3 | 289 | |||
AK293353.1 | - | 4 | 503 | |||
AK310555.1 | - | 3 | 273 | |||
AL832270.1 | - | 3 | 255 | |||
BC045629.1 | - | 3 | 280 | |||
BC130518.1 | - | 3 | 269 | 40146549 | ||
BC143929.1 | - | 3 | 269 | 9052447 | ||
BU532484.1 | - | 3 | 294 | |||
DB147029.1 | - | 3 | 291 | |||
HM991479.1 | - | 3 | 291 | |||
HM991480.1 | - | 3 | 287 | |||
KC736820.1 | - | 2 | 190 |