Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAMTS10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030957.3 | NP_112219.3 | 10, 11 | 1504 | ||
XM_006722917.3 | XP_006722980.1 | 9, 10 | 1350 | |||
XM_017027338.1 | XP_016882827.1 | 9, 10 | 1360 | |||
GenBank mRNA | AB209515.1 | - | 9, 10 | 1340 | ||
AF163762.1 | - | 7, 8 | 1120 | |||
CD632974.1 | - | 4, 5 | 322 |