Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RNF146 SIRNA Assays ›
Gene Symbol:
RNF146
Gene Name:
ring finger protein 146
Gene Aliases:
-
Chromosome Location:
Chr.6: 127266340 - 127289893 on Build GRCh38
UniGene ID:
Hs.267120
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s37821
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RNF146
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242844.1
NP_001229773.1
5
1263
NM_001242845.1
NP_001229774.1
5
1112
NM_001242846.1
NP_001229775.1
5
1025
NM_001242847.1
NP_001229776.1
4
1154
NM_001242848.1
NP_001229777.1
4
884
NM_001242849.1
NP_001229778.1
3
1026
NM_001242850.1
NP_001229779.1
3
756
NM_001242851.1
NP_001229780.1
3
696
NM_001242852.1
NP_001229781.1
5
1243
NM_030963.3
NP_112225.2
5
993
XM_006715571.3
XP_006715634.1
5
1693
XM_011536161.2
XP_011534463.1
6
1580
XM_011536162.2
XP_011534464.1
6
1632
XM_011536163.2
XP_011534465.1
6
1612
XM_011536164.2
XP_011534466.1
5
1471
XM_017011336.1
XP_016866825.1
3
1373
XM_017011337.1
XP_016866826.1
5
4614
XM_017011338.1
XP_016866827.1
7
1721
XM_017011339.1
XP_016866828.1
6
1750
XM_017011340.1
XP_016866829.1
6
1802
XM_017011341.1
XP_016866830.1
4
1501
XM_017011342.1
XP_016866831.1
5
1642
XM_017011343.1
XP_016866832.1
6
1751
GenBank mRNA
AJ315122.1
-
1
558
AJ420549.1
-
2
628
AK027436.1
-
2
697
AK027558.1
-
3
745
AK027776.1
-
1
568
AL136829.1
-
5
956
BC008235.1
-
2
645
3868592
CR533514.1
-
1
557
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
Wnt signaling pathway
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
protein autoubiquitination
protein K48-linked ubiquitination
positive regulation of canonical Wnt signaling pathway
ubiquitin-protein transferase activity
protein binding
zinc ion binding
ligase activity
ubiquitin protein ligase activity
poly-ADP-D-ribose binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-vpmkj:80/100.66.78.144:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline