Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM14B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127711.2 | NP_001121183.1 | 3, 4 | 277 | ||
NM_001286484.1 | NP_001273413.1 | 3, 4 | 302 | |||
NM_001286488.1 | NP_001273417.1 | 4, 5 | 379 | |||
NM_001286489.1 | NP_001273418.1 | 3, 4 | 277 | |||
NM_030969.4 | NP_112231.3 | 4, 5 | 379 | |||
GenBank mRNA | AK291883.1 | - | 4, 5 | 333 | ||
AK294152.1 | - | 4, 5 | 333 | |||
BC001033.1 | - | 4, 5 | 321 | 3139565 | ||
BC007080.1 | - | 4, 5 | 333 | 4066217 | ||
BC013913.1 | - | 4, 5 | 341 | 3685635 | ||
BC071660.1 | - | 4, 5 | 323 | 4473732 | ||
BG432075.1 | - | 4, 5 | 308 | |||
BP249229.1 | - | 3, 4 | 231 | |||
BQ056988.1 | - | 3, 4 | 230 |