Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYB561 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017916.1 | NP_001017916.1 | 3 | 524 | ||
NM_001017917.1 | NP_001017917.1 | 3 | 357 | |||
NM_001915.3 | NP_001906.3 | 3 | 354 | |||
XM_005257091.1 | XP_005257148.1 | 3 | 299 | |||
XM_017024260.1 | XP_016879749.1 | 3 | 396 | |||
XM_017024261.1 | XP_016879750.1 | 2 | 237 | |||
XM_017024262.1 | XP_016879751.1 | 3 | 589 | |||
GenBank mRNA | AK095244.1 | - | 3 | 524 | ||
AK294403.1 | - | 3 | 285 | |||
AK294630.1 | - | 3 | 508 | |||
AK301541.1 | - | 3 | 284 | |||
AK303177.1 | - | 3 | 223 | |||
AK316606.1 | - | 3 | 299 | |||
BC000021.1 | - | 3 | 385 | 3509626 | ||
BC002976.1 | - | 3 | 311 | 3535771 | ||
BC091485.1 | - | 3 | 325 | 6188259 | ||
BF206825.1 | - | 359 | ||||
BM552101.1 | - | 3 | 354 | |||
BT007096.1 | - | 2 | 224 |