Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAS1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170649.1 | NP_001164120.1 | 5 | 845 | ||
NM_001170650.1 | NP_001164121.1 | 4 | 719 | |||
NM_031206.4 | NP_112483.1 | 5 | 845 | |||
XM_005262301.1 | XP_005262358.1 | 5 | 830 | |||
XM_005262304.1 | XP_005262361.1 | 5 | 830 | |||
XM_005262305.3 | XP_005262362.1 | 5 | 830 | |||
XM_005262306.3 | XP_005262363.1 | 5 | 830 | |||
XM_011531045.1 | XP_011529347.1 | 4 | 694 | |||
XM_011531046.1 | XP_011529348.1 | 5 | 830 | |||
XM_017029877.1 | XP_016885366.1 | 5 | 830 | |||
XM_017029878.1 | XP_016885367.1 | 5 | 830 | |||
XM_017029879.1 | XP_016885368.1 | 5 | 830 | |||
XM_017029880.1 | XP_016885369.1 | 5 | 830 | |||
GenBank mRNA | AF116730.1 | - | 5 | 783 | ||
AK022587.1 | - | 5 | 795 | |||
AK074087.1 | - | 5 | 814 | |||
AK097383.1 | - | 4 | 669 | |||
AK225059.1 | - | 5 | 778 | |||
BC014545.1 | - | 5 | 829 | 4079376 | ||
BC018610.1 | - | 5 | 793 | 3140243 | ||
BC019302.1 | - | 5 | 790 | 2822893 |