Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF93 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031218.3 | NP_112495.2 | 4 | 1901 | ||
GenBank mRNA | AK096342.1 | - | 5 | 1774 | ||
AK295921.1 | - | 1 | 1233 | |||
BC136718.1 | - | 4 | 1753 | 9020708 | ||
BC136719.1 | - | 4 | 1753 | 9020709 | ||
BU165449.1 | - | 1 | 206 |