Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF93 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031218.3 | NP_112495.2 | 4 | 1117 | ||
GenBank mRNA | AK022550.1 | - | 1 | 24 | ||
AK096342.1 | - | 4 | 1074 | |||
AK295921.1 | - | 1 | 449 | |||
BC039001.1 | - | 1 | 93 | |||
BC136718.1 | - | 4 | 969 | 9020708 | ||
BC136719.1 | - | 4 | 969 | 9020709 |