Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYBB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000397.3 | NP_000388.2 | 2 | 159 | ||
GenBank mRNA | AK289753.1 | - | 2 | 144 | ||
AK301476.1 | - | 2 | 112 | |||
AK304033.1 | - | 2 | 119 | |||
AK312929.1 | - | 2 | 119 | |||
BC032720.1 | - | 2 | 145 | 5505170 | ||
DA995835.1 | - | 2 | 159 | |||
KU178009.1 | - | 2 | 98 | |||
KU178010.1 | - | 2 | 98 | |||
X04011.1 | - | 2 | 113 |