Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PITPNM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165966.1 | NP_001159438.1 | 15 | 2129 | ||
NM_031220.3 | NP_112497.2 | 16 | 2237 | |||
XM_011524015.2 | XP_011522317.1 | 16 | 2177 | |||
XM_011524016.2 | XP_011522318.1 | 16 | 2178 | |||
GenBank mRNA | AB209130.1 | - | 10 | 4600 | ||
AF334586.1 | - | 16 | 2234 | |||
AL389994.1 | - | 15 | 1960 | |||
BC020484.1 | - | 7 | 835 | |||
BC128583.1 | - | 16 | 2174 | 40128716 | ||
BC128584.1 | - | 16 | 2174 | 40128724 |