Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCF7L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031283.2 | NP_112573.1 | 10, 11 | 1481 | ||
XM_006712109.2 | XP_006712172.1 | 10, 11 | 1274 | |||
GenBank mRNA | AB031046.1 | - | 10, 11 | 1330 | ||
BC058894.1 | - | 10, 11 | 1490 | 6141641 |