Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADPGK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031284.4 | NP_112574.3 | 7 | 1071 | ||
NR_023318.1 | 7 | 1284 | ||||
NR_023319.1 | 5 | 785 | ||||
XM_006720712.3 | XP_006720775.1 | 7 | 1074 | |||
XM_006720713.3 | XP_006720776.1 | 7 | 1235 | |||
XM_011522102.2 | XP_011520404.1 | 4 | 657 | |||
XM_011522103.2 | XP_011520405.1 | 4 | 654 | |||
XM_011522104.2 | XP_011520406.1 | 8 | 1144 | |||
XM_017022664.1 | XP_016878153.1 | 3 | 1015 | |||
GenBank mRNA | AF204270.1 | - | 6 | 663 | ||
AK055526.1 | - | 5 | 776 | |||
AK057651.1 | - | 1 | 1003 | |||
AK075560.1 | - | 7 | 1006 | |||
AK293282.1 | - | 7 | 766 | |||
AK294391.1 | - | 6 | 811 | |||
AL136873.1 | - | 4 | 334 | |||
AL832254.1 | - | 2 | 3092 | |||
BC006112.1 | - | 7 | 1016 | 3347312 | ||
BC044589.1 | - | 7 | 1230 | |||
CR533447.1 | - | 3 | 155 |