Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080554.2 | NP_001074023.1 | 5 | 869 | ||
NM_001080555.2 | NP_001074024.1 | 5 | 745 | |||
NM_001206842.1 | NP_001193771.1 | 4 | 676 | |||
NM_031289.3 | NP_112579.2 | 4 | 776 | |||
NM_153823.3 | NP_722545.2 | 4 | 652 | |||
XM_005253493.2 | XP_005253550.1 | 5 | 799 | |||
XM_005253495.1 | XP_005253552.1 | 5 | 923 | |||
XM_011520858.1 | XP_011519160.1 | 5 | 799 | |||
XM_011520859.1 | XP_011519161.1 | 5 | 716 | |||
XM_011520860.1 | XP_011519162.1 | 5 | 923 | |||
XM_011520861.1 | XP_011519163.1 | 4 | 700 | |||
XM_011520862.1 | XP_011519164.1 | 5 | 840 | |||
XM_011520863.1 | XP_011519165.1 | 4 | 824 | |||
GenBank mRNA | AB593019.1 | - | 4 | 648 | ||
AB593020.1 | - | 4 | 524 | |||
AK027894.1 | - | 4 | 745 | |||
AK058045.1 | - | 5 | 818 | |||
AK075288.1 | - | 4 | 646 | |||
AK075322.1 | - | 4 | 522 | |||
AY358513.1 | - | 4 | 630 | |||
BC001796.1 | - | 4 | 722 | 3353821 | ||
BC033854.1 | - | 4 | 619 | 5164943 | ||
BI831369.1 | - | 5 | 690 |