Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080554.2 | NP_001074023.1 | 5 | 823 | ||
NM_001080555.2 | NP_001074024.1 | 5 | 699 | |||
NM_001206842.1 | NP_001193771.1 | 4 | 630 | |||
NM_031289.3 | NP_112579.2 | 4 | 730 | |||
NM_153823.3 | NP_722545.2 | 4 | 606 | |||
XM_005253493.2 | XP_005253550.1 | 5 | 753 | |||
XM_005253495.1 | XP_005253552.1 | 5 | 877 | |||
XM_011520858.1 | XP_011519160.1 | 5 | 753 | |||
XM_011520859.1 | XP_011519161.1 | 5 | 670 | |||
XM_011520860.1 | XP_011519162.1 | 5 | 877 | |||
XM_011520861.1 | XP_011519163.1 | 4 | 654 | |||
XM_011520862.1 | XP_011519164.1 | 5 | 794 | |||
XM_011520863.1 | XP_011519165.1 | 4 | 778 | |||
GenBank mRNA | AB593019.1 | - | 4 | 602 | ||
AB593020.1 | - | 4 | 478 | |||
AK027894.1 | - | 4 | 699 | |||
AK058045.1 | - | 5 | 772 | |||
AK075322.1 | - | 4 | 476 | |||
AY358513.1 | - | 4 | 584 | |||
BC001796.1 | - | 4 | 676 | 3353821 | ||
BC033854.1 | - | 4 | 573 | 5164943 | ||
BI831369.1 | - | 5 | 644 |