Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDCA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297602.1 | NP_001284531.1 | 3 | 1125 | ||
NM_001297603.1 | NP_001284532.1 | 3 | 1125 | |||
NM_001297604.1 | NP_001284533.1 | 3 | 1125 | |||
NM_031299.5 | NP_112589.1 | 3 | 1125 | |||
NR_123727.1 | 3 | 1125 | ||||
GenBank mRNA | AK092246.1 | - | 3 | 328 | ||
AK291421.1 | - | 3 | 328 | |||
AK312843.1 | - | 3 | 345 | |||
BC002551.2 | - | 3 | 320 | 3138951 | ||
BM911472.1 | - | 3 | 310 | |||
BX099267.1 | - | 2 | 293 | |||
CD106434.1 | - | 3 | 589 | |||
DA735127.1 | - | 3 | 318 |