Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031423.3 | NP_113611.2 | 6 | 559 | ||
NM_145697.2 | NP_663735.2 | 6 | 705 | |||
XM_011510036.1 | XP_011508338.1 | 6 | 501 | |||
GenBank mRNA | AB050577.1 | - | 6 | 620 | ||
AF326731.1 | - | 6 | 536 | |||
AK093348.1 | - | 6 | 684 | |||
BC008489.1 | - | 6 | 492 | 4134468 |