Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201366.1 | NP_001188295.1 | 8, 9 | 741 | ||
NM_031427.3 | NP_113615.2 | 7, 8 | 695 | |||
XM_017021679.1 | XP_016877168.1 | 8, 9 | 729 | |||
GenBank mRNA | AF542071.1 | - | 7, 8 | 571 | ||
AK026653.1 | - | 4, 5 | 353 | |||
AK315392.1 | - | 7, 8 | 590 | |||
AL833654.1 | - | 6, 7 | 451 | |||
BC005343.1 | - | 7, 8 | 571 | 3930213 | ||
BJ996719.1 | - | 3, 4 | 291 |