Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASSF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182663.3 | NP_872604.1 | 4 | 1004 | ||
NM_182664.3 | NP_872605.1 | 4 | 1004 | |||
NM_182665.3 | NP_872606.1 | 3 | 882 | |||
GenBank mRNA | AB209145.1 | - | 4 | 939 | ||
AF445801.1 | - | 3 | 882 | |||
AK056568.1 | - | 1 | 153 | |||
AK074115.1 | - | 2 | 349 | |||
AK289861.1 | - | 4 | 1004 | |||
AK291268.1 | - | 4 | 1002 | |||
AK292039.1 | - | 4 | 740 | |||
AL832784.1 | - | 3 | 782 | |||
AY062002.1 | - | 4 | 988 | |||
AY062003.1 | - | 4 | 988 | |||
AY216268.1 | - | 3 | 875 | |||
AY261332.1 | - | 3 | 782 | |||
BC004270.1 | - | 4 | 988 | 3613993 | ||
BC007203.1 | - | 4 | 988 | 2967057 | ||
BC042651.1 | - | 3 | 862 | 4342962 |