Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOX7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031439.3 | NP_113627.1 | 2 | 1140 | ||
GenBank mRNA | AJ409320.1 | - | 2 | 999 | ||
AK055556.1 | - | 2 | 1078 | |||
AK223574.1 | - | 2 | 1078 | |||
BC004299.1 | - | 2 | 1067 | |||
BC071947.1 | - | 2 | 1066 | 6459570 | ||
BT006693.1 | - | 2 | 999 |