Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP1A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000761.4 | NP_000752.2 | 2 | 725 | ||
GenBank mRNA | AF182274.1 | - | 1 | 669 | ||
BC067424.1 | - | 2 | 705 | 7002123 | ||
BC067425.1 | - | 2 | 705 | 7002124 | ||
BC067426.1 | - | 2 | 705 | 7002125 | ||
BC067427.1 | - | 2 | 705 | 7002126 | ||
BC067428.1 | - | 2 | 705 | 7002128 | ||
BC067429.1 | - | 2 | 705 | 7002129 | ||
M55053.1 | - | 1 | 669 | |||
Z00036.1 | - | 1 | 669 |