Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031457.1 | NP_113645.1 | 6, 7 | 855 | ||
GenBank mRNA | AF237905.1 | - | 6, 7 | 855 | ||
AF350504.1 | - | 6, 7 | 697 | |||
BC022895.1 | - | 6, 7 | 780 | 4691480 |