Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146102.1 | NP_001139574.1 | 8 | 1895 | ||
NM_001146103.1 | NP_001139575.1 | 8 | 1972 | |||
NM_001146104.1 | NP_001139576.1 | 8 | 1971 | |||
NM_001146105.1 | NP_001139577.1 | 8 | 1790 | |||
NM_001146106.1 | NP_001139578.1 | 8 | 1971 | |||
NM_031458.2 | NP_113646.2 | 8 | 2077 | |||
XM_005247820.2 | XP_005247877.1 | 8 | 2001 | |||
XM_011513218.1 | XP_011511520.1 | 8 | 1867 | |||
XM_017007304.1 | XP_016862793.1 | 8 | 1885 | |||
XM_017007305.1 | XP_016862794.1 | 8 | 1780 | |||
XM_017007306.1 | XP_016862795.1 | 8 | 1861 | |||
GenBank mRNA | AB209742.1 | - | 4 | 594 | ||
AF307338.1 | - | 8 | 1978 | |||
AF307339.1 | - | 8 | 1873 | |||
AK123412.1 | - | 2 | 833 | |||
AK130147.1 | - | 3 | 426 | |||
AK292959.1 | - | 8 | 1750 | |||
AK299107.1 | - | 8 | 1864 | |||
AK313494.1 | - | 8 | 1864 | |||
AL713679.1 | - | 1 | 178 | |||
AL832929.1 | - | 1 | 110 | |||
BC017463.1 | - | 8 | 1939 | |||
BC039580.1 | - | 7 | 1748 | 5416494 | ||
BP259272.1 | - | 3 | 349 | |||
BX648869.1 | - | 6 | 617 |