Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS6KL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031464.4 | NP_113652.2 | 11 | 2549 | ||
XM_017021685.1 | XP_016877174.1 | 13 | 1987 | |||
XM_017021686.1 | XP_016877175.1 | 11 | 2526 | |||
XM_017021689.1 | XP_016877178.1 | 10 | 1543 | |||
XM_017021690.1 | XP_016877179.1 | 9 | 1901 | |||
XM_017021692.1 | XP_016877181.1 | 9 | 1453 | |||
GenBank mRNA | AK093053.1 | - | 4 | 1201 | ||
AK131532.1 | - | 3 | 2583 | |||
AK226137.1 | - | 9 | 2810 | |||
AL390152.1 | - | 3 | 290 | |||
BC004540.2 | - | 11 | 1792 | 3945902 |