Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAPPC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160372.3 | NP_001153844.1 | 15 | 2327 | ||
NM_001321646.1 | NP_001308575.1 | 14 | 2300 | |||
NM_031466.7 | NP_113654.4 | 15 | 2502 | |||
XM_011517326.2 | XP_011515628.1 | 14 | 2475 | |||
XM_011517328.2 | XP_011515630.1 | 15 | 2502 | |||
XM_011517330.2 | XP_011515632.1 | 7 | 867 | |||
XM_017013893.1 | XP_016869382.1 | 15 | 2502 | |||
XM_017013894.1 | XP_016869383.1 | 8 | 1203 | |||
GenBank mRNA | AB067469.1 | - | 15 | 2277 | ||
AK026394.1 | - | 15 | 2319 | |||
AK027689.1 | - | 4 | 427 | |||
AK128755.1 | - | 15 | 2503 | |||
AK308815.1 | - | 4 | 598 | |||
AL833973.1 | - | 13 | 1602 | |||
AY190606.1 | - | 13 | 1581 | |||
BC003650.2 | - | 1 | 46 | |||
BC006206.2 | - | 13 | 1723 | 3538806 |