Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017440.2 | NP_001017440.1 | 2, 3 | 643 | ||
NM_031468.3 | NP_113656.2 | 3, 4 | 633 | |||
XM_011516594.2 | XP_011514896.1 | 2, 3 | 485 | |||
XM_011516595.1 | XP_011514897.1 | 2, 3 | 233 | |||
XM_011516596.2 | XP_011514898.1 | 3, 4 | 1128 | |||
XM_011516597.2 | XP_011514899.1 | 2, 3 | 1061 | |||
XM_011516599.1 | XP_011514901.1 | 2, 3 | 432 | |||
XM_017012675.1 | XP_016868164.1 | 4, 5 | 988 | |||
XM_017012676.1 | XP_016868165.1 | 3, 4 | 770 | |||
XM_017012677.1 | XP_016868166.1 | 2, 3 | 283 | |||
XM_017012678.1 | XP_016868167.1 | 3, 4 | 745 | |||
XM_017012679.1 | XP_016868168.1 | 2, 3 | 1098 | |||
XM_017012680.1 | XP_016868169.1 | 2, 3 | 397 | |||
XM_017012681.1 | XP_016868170.1 | 2, 3 | 260 | |||
XM_017012682.1 | XP_016868171.1 | 2, 3 | 810 | |||
XM_017012683.1 | XP_016868172.1 | 3, 4 | 594 | |||
GenBank mRNA | AF282250.1 | - | 3, 4 | 364 | ||
AK123862.1 | - | 2, 3 | 178 | |||
AK313054.1 | - | 2, 3 | 233 | |||
AY007302.1 | - | 2, 3 | 291 | |||
AY726595.1 | - | 1, 2 | 119 | |||
BC020200.1 | - | 2, 3 | 415 | 3944302 | ||
BG718538.1 | - | 3, 4 | 638 |