Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3BGRL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031469.3 | NP_113657.1 | 2, 3 | 584 | ||
XM_011536165.2 | XP_011534467.1 | 2, 3 | 558 | |||
XM_017011344.1 | XP_016866833.1 | 4, 5 | 5624 | |||
GenBank mRNA | AF340151.1 | - | 2, 3 | 406 | ||
AJ297972.1 | - | 2, 3 | 227 | |||
AK074506.1 | - | 2, 3 | 386 | |||
AK307145.1 | - | 2, 3 | 644 | |||
AK311757.1 | - | 2, 3 | 392 | |||
BC040489.1 | - | 2, 3 | 392 | 5286092 | ||
BC052987.1 | - | 2, 3 | 384 | 5503583 | ||
BC060799.1 | - | 2, 3 | 360 | 30330699 | ||
BC070059.1 | - | 2, 3 | 360 | 30346325 | ||
BC109043.2 | - | 2, 3 | 394 | 40028800 | ||
BC109044.2 | - | 2, 3 | 394 | 40028802 |