Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM57B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318504.1 | NP_001305433.1 | 5 | 519 | ||
NM_031478.5 | NP_113666.2 | 5 | 957 | |||
XM_005255613.3 | XP_005255670.1 | 9 | 1165 | |||
XM_005255614.3 | XP_005255671.1 | 8 | 1105 | |||
XM_005255615.2 | XP_005255672.1 | 8 | 1302 | |||
XM_017023749.1 | XP_016879238.1 | 6 | 1149 | |||
XM_017023750.1 | XP_016879239.1 | 9 | 1362 | |||
XM_017023751.1 | XP_016879240.1 | 10 | 1258 | |||
XM_017023752.1 | XP_016879241.1 | 9 | 1161 | |||
XM_017023753.1 | XP_016879242.1 | 7 | 1241 | |||
XM_017023754.1 | XP_016879243.1 | 6 | 663 | |||
GenBank mRNA | AB593089.1 | - | 5 | 897 | ||
AB593090.1 | - | 5 | 895 | |||
AF370365.1 | - | 5 | 909 | |||
AL136777.1 | - | 5 | 519 | |||
BC007892.2 | - | 5 | 675 | 4126084 |