Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITCH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257137.2 | NP_001244066.1 | 11 | 1238 | ||
NM_001257138.2 | NP_001244067.1 | 9 | 973 | |||
NM_001324197.1 | NP_001311126.1 | 11 | 1259 | |||
NM_001324198.1 | NP_001311127.1 | 10 | 1136 | |||
NM_031483.6 | NP_113671.3 | 10 | 1115 | |||
XM_005260570.1 | XP_005260627.1 | 9 | 1031 | |||
XM_011529079.1 | XP_011527381.1 | 9 | 1051 | |||
XM_017028089.1 | XP_016883578.1 | 10 | 1071 | |||
XM_017028090.1 | XP_016883579.1 | 11 | 1191 | |||
XM_017028091.1 | XP_016883580.1 | 9 | 986 | |||
XM_017028092.1 | XP_016883581.1 | 11 | 1191 | |||
GenBank mRNA | AB056663.2 | - | 10 | 1025 | ||
AB209747.1 | - | 2 | 132 | |||
AF038564.1 | - | 5 | 533 | |||
AF095745.1 | - | 8 | 908 | |||
AK297809.1 | - | 10 | 933 | |||
AK304090.1 | - | 9 | 932 | |||
AK315212.1 | - | 10 | 1074 | |||
BC011571.1 | - | 11 | 1161 | 4560366 |