Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LONP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300948.1 | NP_001287877.1 | 8 | 1511 | ||
NM_031490.3 | NP_113678.2 | 9 | 1643 | |||
XM_017023755.1 | XP_016879244.1 | 9 | 1510 | |||
XM_017023756.1 | XP_016879245.1 | 8 | 1376 | |||
GenBank mRNA | AJ548761.1 | - | 9 | 1510 | ||
AK027544.1 | - | 9 | 1476 | |||
AK027666.1 | - | 5 | 678 | |||
AK074775.1 | - | 9 | 1508 | |||
AK294779.1 | - | 8 | 1323 | |||
AL834201.1 | - | 8 | 1009 | |||
BC093910.1 | - | 9 | 1477 | 7939755 | ||
BC093912.1 | - | 9 | 1477 | 7939757 | ||
BC110434.1 | - | 5 | 659 | |||
BC143246.1 | - | 8 | 1345 | 9051747 |