Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRTAP4-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031854.2 | NP_114060.1 | 1 | 907 | ||
GenBank mRNA | AJ406943.1 | - | 1 | 907 | ||
BC004180.2 | - | 1 | 887 | 2959821 | ||
BC004212.2 | - | 1 | 887 | 2959821 | ||
BC101159.1 | - | 1 | 899 | 40015747 | ||
BC101160.1 | - | 1 | 899 | 40015748 | ||
BC101161.1 | - | 2 | 434 | |||
BC101162.1 | - | 1 | 899 | 40015750 |