Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP2B6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000767.4 | NP_000758.1 | 3 | 390 | ||
XM_005258569.4 | XP_005258626.1 | 3 | 486 | |||
XM_006723050.3 | XP_006723113.1 | 3 | 486 | |||
XM_011526546.2 | XP_011524848.1 | 3 | 489 | |||
XM_011526547.2 | XP_011524849.1 | 3 | 487 | |||
XM_011526548.2 | XP_011524850.1 | 3 | 483 | |||
XM_011526550.2 | XP_011524852.1 | 2 | 278 | |||
GenBank mRNA | AF182277.1 | - | 3 | 385 | ||
AK301620.1 | - | 2 | 267 | |||
BC067430.1 | - | 3 | 390 | 7002137 | ||
BC067431.1 | - | 3 | 390 | 7002142 | ||
M29874.1 | - | 3 | 389 | |||
X13494.1 | - | 3 | 322 |