Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARMC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161009.2 | NP_001154481.1 | 4, 5 | 993 | ||
NM_031905.4 | NP_114111.2 | 5, 6 | 1098 | |||
XM_011516601.2 | XP_011514903.1 | 4, 5 | 1171 | |||
XM_011516602.2 | XP_011514904.1 | 4, 5 | 532 | |||
XM_017012684.1 | XP_016868173.1 | 6, 7 | 1279 | |||
XM_017012687.1 | XP_016868176.1 | 4, 5 | 511 | |||
GenBank mRNA | AK075500.1 | - | 5, 6 | 839 | ||
AK225082.1 | - | 4, 5 | 726 | |||
AK291818.1 | - | 4, 5 | 721 | |||
AY150853.1 | - | 4, 5 | 993 | |||
AY150854.1 | - | 5, 6 | 1098 | |||
BC003586.1 | - | 4, 5 | 713 | 3357276 |